Order Entry
Export
ContactUsLinkComponent
Magnetic roll holder, PIG®
Magnetic roll holder, PIG®
Catalog # NEWP57701
Supplier:  NEW PIG
Magnetic roll holder, PIG®
Catalog # NEWP57701
Supplier:  NEW PIG

Specifications

  • Description:
    Roll holder, powder coated steel, 2 kg
  • W×D×H:
    152×81×457 mm
  • Pk:
    1

Specifications

About this item

Magnetic roll holder helps make accessing and storing PIG® Absorbent mats easy and convenient.

  • Holds one 38 cm × 15 m PIG® mat roll for easy one-hand dispensing
  • Durable, powder-coated steel resists chips, rust and corrosion
  • Powerful magnets keep roll holder firmly in place during dispensing; can be removed and relocated in seconds
  • Ideal for attaching to tool boxes, car lifts, support beams or other metal surfaces

Magnetic holder attaches to any metal surface to keep your PIG® Mat roll right where you need it for quick cleanups.