



Petroleum spirit, 100…140 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:64742-49-0
:64742-82-1
MDL Number:MFCD00081849
Boiling Pt:100…140 °C (1013 hPa)
Storage Temperature:Ambient
UN:1268
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
Specification Test Results
- Substances coloured by H₂SO₄:Passes test
- Acidity:Max. 0.0003 meq/g
- Density (20/4):0.690 - 0.750
- Distillation (min. 90 %):100 - 140 °C
- Evaporation residue:Max. 20 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC23857.320 spec_VWRC23857.320 spec_VWRC23857.460 spec_VWRC23857.460