Thermal transfer printable labels, series R6400
Supplier: Brady
Certificates
About this item
Polypropylene (B-7425) material is ideal for general purpose barcode applications and withstands liquid nitrogen, autoclave and other harsh environments.
- Die-cut labels are on a roll
- General identification, laboratory labelling and temperature indicating labelling applications
- Service temperature: –80 to 70 °C
- This R6400 series has matt finish
- Thickness: 0,102 mm
- Material with autoclavable and cryogenic storage properties
This node contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates. This node contains only supplies sold with the ribbon seperate, it is not in the same enclosure as the tape/sleeves.
For freezer or liquid nitrogen applications, B-7425 must be applied prior to being frozen.