

Tripotassium orthophosphate trihydrate ≥98.0%, AnalaR NORMAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:22763-03-7
:
Formula:K₃O₄P·3H₂O
MDL Number:MFCD00151709
:266.31 g/mol
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay :≥ 98.0 %
- Appearance :White hygroscopic powder
- Identification:Passes test
- di-Potassium hydrogen phosphate :≤ 2.0 %
- Free alkali (as KOH) :≤ 1.0 %
- Heavy metals (as Pb) :≤ 20 ppm
- Total N (Nitrogen) :≤ 10 ppm
- Cl (Chloride) :≤ 30 ppm
- As (Arsenic) :≤ 1 ppm
- Fe (Iron) :≤ 10 ppm
- Na (Sodium) :≤ 0.5 %
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC26937.290 spec_VWRC26937.460
Product specification
VWR Chemicals Big Packs and Bulks