Order Entry
Export
ContactUsLinkComponent
illustra™ blood genomicPrep mini spin kit
illustra™ blood genomicPrep mini spin kit
Supplier:  Cytiva
Certificates
undefined
illustra™ blood genomicPrep mini spin kit
Supplier:  Cytiva

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Blood
  • Target molecule:
    DNA
  • Description:
    illustra™ blood genomicPrep Mini Spin Kit

Specifications