Order Entry
Export
Orders LinkContactUsLinkComponent
Flame-retardant overalls, FR80

Flame-retardant overalls, FR80

Supplier:   Portwest
Flame-retardant overalls, FR80
Supplier:  Portwest
Certificates
Flame-retardant overalls, FR80

7 Options Available Below

Product Details & Documents

These overalls are perfect for the demands of the petrochemical industry. The highly technical fabric provides flame, heat, iron splash, chemical, electric arc and anti-static protection. The high collar and studded flaps on each pocket provide maximum protection.

  • Protection against radiant, convective and contact heat
  • Class 2 welding protection
  • Knee pad pockets to facilitate knee pads
  • Triple-stitched seams for extra durability
  • 50+ UPF rated fabric to block 98% of UV rays
  • Nine pockets for ample storage
  • Radio loops for easy clipping of a radio

Colour: Navy blue

Weight: 345 g/m²

Certifications: EN 1149 -5, EN ISO 11612 (A1+A2 B1, C1, E3, F1), EN ISO 11611 (Class 1 A1+A2), IEC 61482-2 (Class 1), EN 13034 (Type 6), ASTM F1959/F1959M-12

Specifications for Product Family

Specifications

  • Catalog No:
  • PRTWFR80NARS
  • PRTWFR80NARM
  • PRTWFR80NARL
  • PRTWFR80NARXL
  • PRTWFR80NARXXL
  • PRTWFR80NARXXXL
  • PRTWFR80NAR4XL
  • Model:
  • Bizflame FR80
  • Bizflame FR80
  • Bizflame FR80
  • Bizflame FR80
  • Bizflame FR80
  • Bizflame FR80
  • Bizflame FR80
  • Garment application:
  • Chemical protection|Industrial
  • Chemical protection|Industrial
  • Chemical protection|Industrial
  • Chemical protection|Industrial
  • Chemical protection|Industrial
  • Chemical protection|Industrial
  • Chemical protection|Industrial
  • Risk area:
  • Heat|Flame-retardant|Anti-static
  • Heat|Flame-retardant|Anti-static
  • Heat|Flame-retardant|Anti-static
  • Heat|Flame-retardant|Anti-static
  • Heat|Flame-retardant|Anti-static
  • Heat|Flame-retardant|Anti-static
  • Heat|Flame-retardant|Anti-static
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Weight:
  • 345 g/m²
  • 345 g/m²
  • 345 g/m²
  • 345 g/m²
  • 345 g/m²
  • 345 g/m²
  • 345 g/m²

Specifications

Product Family Options

Product Information

  • Colour
    Clothing Size
    Availability
    Price
  • Dark blue
    S
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 157,00 €
    157,00 €
    1 * 1 items
       157,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    PRTWFR80NARS
    Model
    Bizflame FR80
    Garment application
    Chemical protection|Industrial
    Risk area
    Heat|Flame-retardant|Anti-static
    Gender
    Unisex
    Sleeve or leg length
    Long
    Weight
    345 g/m²
  • Dark blue
    M
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 157,00 €
    157,00 €
    1 * 1 items
       157,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    PRTWFR80NARM
    Model
    Bizflame FR80
    Garment application
    Chemical protection|Industrial
    Risk area
    Heat|Flame-retardant|Anti-static
    Gender
    Unisex
    Sleeve or leg length
    Long
    Weight
    345 g/m²
  • Dark blue
    L
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 157,00 €
    157,00 €
    1 * 1 items
       157,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    PRTWFR80NARL
    Model
    Bizflame FR80
    Garment application
    Chemical protection|Industrial
    Risk area
    Heat|Flame-retardant|Anti-static
    Gender
    Unisex
    Sleeve or leg length
    Long
    Weight
    345 g/m²
  • Dark blue
    XL
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 157,00 €
    157,00 €
    1 * 1 items
       157,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    PRTWFR80NARXL
    Model
    Bizflame FR80
    Garment application
    Chemical protection|Industrial
    Risk area
    Heat|Flame-retardant|Anti-static
    Gender
    Unisex
    Sleeve or leg length
    Long
    Weight
    345 g/m²
  • Dark blue
    XXL
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 157,00 €
    157,00 €
    1 * 1 items
       157,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    PRTWFR80NARXXL
    Model
    Bizflame FR80
    Garment application
    Chemical protection|Industrial
    Risk area
    Heat|Flame-retardant|Anti-static
    Gender
    Unisex
    Sleeve or leg length
    Long
    Weight
    345 g/m²
  • Dark blue
    3XL
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 157,00 €
    157,00 €
    1 * 1 items
       157,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    PRTWFR80NARXXXL
    Model
    Bizflame FR80
    Garment application
    Chemical protection|Industrial
    Risk area
    Heat|Flame-retardant|Anti-static
    Gender
    Unisex
    Sleeve or leg length
    Long
    Weight
    345 g/m²
  • Dark blue
    4XL
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 157,00 €
    157,00 €
    1 * 1 items
       157,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    PRTWFR80NAR4XL
    Model
    Bizflame FR80
    Garment application
    Chemical protection|Industrial
    Risk area
    Heat|Flame-retardant|Anti-static
    Gender
    Unisex
    Sleeve or leg length
    Long
    Weight
    345 g/m²