General purpose gloves, black, A400
Supplier: Portwest
Certificates
About this item
This PVC fully dipped working glove with knitted wrist and cotton interlock lining has a smooth finish with excellent abrasion resistance.
- 100% breathable seamless liner
- Superb abrasion and tear resistance
- 13 gauge liner for a perfect fit
- Knitted cuff for comfort and warmth
- Fully coated for maximum liquid protection
Protects against oil in warm and humid conditions.
Certifications: AS/NZS 2161 .2; AS/NZS 2161 .3; ANSI/ISEA 105: 2016 CUT Level (A1); EN 388:2016 (4121X)
Packaging: 12 pairs per pack.
Specifications
- Catalog No:
- PRTWA400RERXL
- PRTWA400RERM
- PRTWA400RERL
- PRTWA400BKRXXL
- PRTWA400BKRL
- PRTWA400RERXXL
- PRTWA400BKRXL
- PRTWA400BKRM
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Cotton
- Cotton
- Cotton
- Cotton
- Cotton
- Cotton
- Finish:
- Full PVC coating
- Full PVC coating
- Full PVC coating
- Full PVC coating
- Full PVC coating
- Full PVC coating
- Full PVC coating
- Full PVC coating
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)