Orders LinkContactUsLinkComponent
Cut resistant gloves, AP50
Cut resistant gloves, AP50
Supplier:   Portwest
Cut resistant gloves, AP50
Supplier:  Portwest
Certificates
5 Options Available Below
Product Details & Documents
The fully coated Aqua Cut Pro glove offers level D cut resistance.
  • Sandy finish for exceptional grip in water, grease or oil
  • Protects against cuts and abrasion
  • Prevents grease, oil and water penetration
  • Seamless 13 gauge liner
  • Retail bag which aids presentation for retail sales
  • ANSI Cut Level A4
The first coat is a smooth nitrile and the second coat is foam nitrile, providing a secure grip and a barrier against contaminants. Elastic wrist cuff provides a secure fit.
Certifications: AS/NZS 2161 .2, AS/NZS 2161 .3, ANSI/ISEA 105 - 2016 CUT Level (A4), EN 420, EN 388:2016 - (4.X.4.4.D)
Specifications for Product Family
Specifications
  • Catalog No:
  • PRTWAP50B8RS
  • PRTWAP50B8RM
  • PRTWAP50B8RL
  • PRTWAP50B8RXL
  • PRTWAP50B8RXXL
  • Model:
  • AP50
  • AP50
  • AP50
  • AP50
  • AP50
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • HPPE
  • HPPE
  • HPPE
  • HPPE
  • HPPE
  • Finish:
  • Fully coated
  • Fully coated
  • Fully coated
  • Fully coated
  • Fully coated
  • Gauge:
  • 13
  • 13
  • 13
  • 13
  • 13
  • Cuff style:
  • Elastic
  • Elastic
  • Elastic
  • Elastic
  • Elastic
  • EN norm:
  • EN 420, EN 388:2016 - (4.X.4.4.D)
  • EN 420, EN 388:2016 - (4.X.4.4.D)
  • EN 420, EN 388:2016 - (4.X.4.4.D)
  • EN 420, EN 388:2016 - (4.X.4.4.D)
  • EN 420, EN 388:2016 - (4.X.4.4.D)
Specifications
Product Family Options
Product Information
  • Glove size
    Colour
    Availability
    Price
  • S
    Blue/black
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR - 151,00 €
    151,00 €
    1 * 12 PAIR
       151,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    PRTWAP50B8RS
    Model
    AP50
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    HPPE
    Finish
    Fully coated
    Gauge
    13
    Cuff style
    Elastic
    EN norm
    EN 420, EN 388:2016 - (4.X.4.4.D)
  • M
    Blue/black
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR - 151,00 €
    151,00 €
    1 * 12 PAIR
       151,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    PRTWAP50B8RM
    Model
    AP50
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    HPPE
    Finish
    Fully coated
    Gauge
    13
    Cuff style
    Elastic
    EN norm
    EN 420, EN 388:2016 - (4.X.4.4.D)
  • L
    Blue/black
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR - 151,00 €
    151,00 €
    1 * 12 PAIR
       151,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    PRTWAP50B8RL
    Model
    AP50
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    HPPE
    Finish
    Fully coated
    Gauge
    13
    Cuff style
    Elastic
    EN norm
    EN 420, EN 388:2016 - (4.X.4.4.D)
  • XL
    Blue/black
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR - 151,00 €
    151,00 €
    1 * 12 PAIR
       151,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    PRTWAP50B8RXL
    Model
    AP50
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    HPPE
    Finish
    Fully coated
    Gauge
    13
    Cuff style
    Elastic
    EN norm
    EN 420, EN 388:2016 - (4.X.4.4.D)
  • XXL
    Blue/black
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR - 151,00 €
    151,00 €
    1 * 12 PAIR
       151,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    PRTWAP50B8RXXL
    Model
    AP50
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    HPPE
    Finish
    Fully coated
    Gauge
    13
    Cuff style
    Elastic
    EN norm
    EN 420, EN 388:2016 - (4.X.4.4.D)