Orders LinkContactUsLinkComponent
Cut resistant gloves, A621
Cut resistant gloves, A621
Supplier:   Portwest
Cut resistant gloves, A621
Supplier:  Portwest
Certificates
Cut resistant gloves, A621
5 Options Available Below
Product Details & Documents
Cut resistance combined with superb grip in oily conditions. ³/₄ nitrile foam dipping absorbs oil keeping hands clean and dry to ensure secure handling.
  • ANSI cut level A3
  • Materials: HDPE, glass fibre, nitrile foam
  • 100% breathable seamless liner
  • 13 gauge liner for a perfect fit
  • Level D cut resistance with abrasion and tear resistance
Nitrile foam coating for excellent grip in wet and dry conditions.
Certifications: Standards: AS/NZS 2161 .2, AS/NZS 2161 .3, AS/NZS 2161 .4, ANSI/ISEA 105: 2016 CUT Level A4, EN388:2016 +A1:2018 - 4.X.4.3.D., EN 407 X.1.X.X.X.X., EN 420.
Specifications for Product Family
Specifications
  • Catalog No:
  • PRTWA621BKRS
  • PRTWA621BKRM
  • PRTWA621BKRL
  • PRTWA621BKRXL
  • PRTWA621BKRXXL
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
Specifications
Product Family Options
Product Information
  • Glove size
    Colour
    Availability
    Price
  • S
    Black/grey
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR - 93,30 €
    93,30 €
    1 * 12 PAIR
       93,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    PRTWA621BKRS
    Ambidextrous / hand-specific
    Hand-specific
  • M
    Black/grey
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR - 93,30 €
    93,30 €
    1 * 12 PAIR
       93,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    PRTWA621BKRM
    Ambidextrous / hand-specific
    Hand-specific
  • L
    Black/grey
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR - 93,30 €
    93,30 €
    1 * 12 PAIR
       93,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    PRTWA621BKRL
    Ambidextrous / hand-specific
    Hand-specific
  • XL
    Black/grey
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR - 93,30 €
    93,30 €
    1 * 12 PAIR
       93,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    PRTWA621BKRXL
    Ambidextrous / hand-specific
    Hand-specific
  • XXL
    Black/grey
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR - 93,30 €
    93,30 €
    1 * 12 PAIR
       93,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    PRTWA621BKRXXL
    Ambidextrous / hand-specific
    Hand-specific