1,3-Dimethyladamantane
Catalog # MOLE25531586-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:164.29 g/mol
Boiling Pt:201.5 °C (1013 hPa)
Melting Pt:–30 °C
Density:0.9016 g/cm³ (20 °C)
EINECS:211-870-8
UN:3295
ADR:3,III
CAS Number:702-79-4
Formula:C₁₂H₂₀
MDL Number:MFCD00074755