
1,3-Dimethyladamantane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:702-79-4
:
Formula:C₁₂H₂₀
MDL Number:MFCD00074755
:164.29 g/mol
Boiling Pt:201 °C
Melting Pt:–30 °C
Density:0.902
Flash Pt:52 °C (126 °F)
Storage Temperature:Ambient
EINECS:211-870-8
UN:3295
ADR:3,III
:GHS02