Order Entry
Export
Orders LinkContactUsLinkComponent
Melamine

Melamine

Glass bottle for solids, 1 kg
Catalog # ROTH6034.1
Supplier:   Roth Carl
Melamine
Catalog # ROTH6034.1
Supplier:  Roth Carl
CAS Number:  108-78-1
Melamine

Glass bottle for solids, 1 kg

CAS Number:  108-78-1
Price
63,90 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  2,4,6-Triamino-s-triazine, 1,3,5-Triazine-2,4,6-triamine, 2,4,6-Triamino-1,3,5-triazine
  • CAS Number:108-78-1
  • Formula:C₃H₆N₆
  • MDL Number:MFCD00006055
  • Molecular Weight:126.12 g/mol
  • Merck Index:13,05834
  • Density:1.57 g/cm³ (20 °C)
  • Flash Pt:>280 °C
  • EINECS:203-615-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications