Order Entry
Export
Orders LinkContactUsLinkComponent
Atrazine, extra pure

Atrazine, extra pure

Glass ampoule, 10 mg
Catalog # 123542H
Health
Environment
Supplier:   VWR Chemicals
Atrazine, extra pure
Catalog # 123542H
Supplier:  VWR Chemicals
CAS Number:  1912-24-9
Atrazine, extra pure
Atrazine, extra pure

Glass ampoule, 10 mg

CAS Number:  1912-24-9
Price
51,30 €
Log in for availability
1 * 10 mg
Product Details & Documents
Synonyms:  2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine
  • CAS Number:1912-24-9
  • Formula:C₈H₁₄ClN₅
  • MDL Number:MFCD00041810
  • Molecular Weight:215.69 g/mol
  • Merck Index:12,00902
  • Boiling Pt:369 °C (1013 hPa)
  • Melting Pt:171…174 °C
  • Density:1.2 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:217-617-8
  • ADR:9,III
  •  :Density: 1.2 g/cm³ (20 °C)
  • UN:3077

Documents

Specifications

Specifications

  • Pack type:
    Glass ampoule
  • Pk:
    10 mg

Specifications