Order Entry
Export
Orders LinkContactUsLinkComponent
2,3-Dimethylthiophene ≥97%

2,3-Dimethylthiophene ≥97%

Flammable
Irritant
2,3-Dimethylthiophene ≥97%
Certificates
2,3-Dimethylthiophene ≥97%
2,3-Dimethylthiophene ≥97%

2 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 632-16-6 Molecular Formula: C6H8S Formula Weight: 112.19 Flash Point: 26° (79°F) Boiling Point: 142° UN No.: 1993 MDL No.: MFCD00130081

  • CAS Number:632-16-6
  • Formula:C₆H₈S
  • MDL Number:MFCD00130081
  • Molecular Weight:112.2 g/mol
  • Boiling Pt:142 °C
  • Melting Pt:–49 °C
  • Density:1.0021 g/cm³ (20 °C)
  • Flash Pt:26 °C (79 °F)
  • Storage Temperature:Ambient
  • EINECS:211-170-2
  • ADR:3,III
  •  :Density: 1,0021 g/cm³ (20 °C)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 90,90 €
    90,90 €
    1 * 1 g
       90,90 €

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 363,00 €
    363,00 €
    1 * 5 g
       363,00 €