3-Hydroxy-2-nitropyridine
Glass bottle for solids, 25 g
Catalog # MOLEM63119725
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 2-Nitropyridin-3-ol, 2-Nitro-3-pyridinol
- CAS Number:15128-82-2
- Formula:C₅H₄N₂O₃
- MDL Number:MFCD00006258
- Molecular Weight:140.1 g/mol
- Melting Pt:70 °C
- EINECS:239-191-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



