Order Entry
Export
ContactUsLinkComponent
3-Hydroxy-2-nitropyridine 97%
Irritant
3-Hydroxy-2-nitropyridine 97%
Certificates
undefined
Irritant
3-Hydroxy-2-nitropyridine 97%

Specification Test Results

  • Appearance:
    Yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    67 to 72 °C
  • GC:
    ≥96.0 %

Specification Test Results

Related Information