
Methylene blue
Catalog # ROTHA514.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
Specifications
About this item
:
:319.86 g/mol
Merck Index:12,06137
Melting Pt:180 °C
Density:1.757 g/cm³ (22 °C)
EINECS:200-515-2
:GHS07
Storage Temperature:Ambient
CAS Number:61-73-4
:MFCD00012111
Formula:C₁₆H₁₈ClN₃S
MDL Number:MFCD00150008