

1,4-Butanesultone 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1633-83-6
:
Formula:C₄H₈O₃S
MDL Number:MFCD00006584
:136.17 g/mol
Boiling Pt:153 °C (19hPa)
Melting Pt:12.5…15 °C
Density:1.331 g/cm³
Flash Pt:154 °C
Storage Temperature:Ambient
EINECS:216-647-9
:GHS07
:GHS08
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4600 to 1.4640 (20°C, 589 nm)
- Water:≤0.1 % (K.F.)
- GC:≥99.0 %
- Titration with TBAH:≤0.1 % 4-Hydoxybutanesulfonic acid
- Chloride (Cl):≤100 ppm