Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dimethyl-4-pyrone
2,6-Dimethyl-4-pyrone
2,6-Dimethyl-4-pyrone
2,6-Dimethyl-4-pyrone
Supplier:
CHEMPUR
Certificates
Expand image
2,6-Dimethyl-4-pyrone
Supplier:
CHEMPUR
About this item
CAS Number:
1004-36-0
:
Formula:
C₇H₈O₂
MDL Number:
MFCD00006579
Boiling Pt:
248 °C (1013 hPa)
Melting Pt:
130…131 °C
Density:
1.17 g/cm³ (25 °C)
:
124.14 g/mol
EINECS:
213-719-1