Order Entry
Export
ContactUsLinkComponent
Zirconium(IV) oxide
Irritant
Zirconium(IV) oxide
Catalog # ROTH9303.1
Supplier:  Roth Carl
CAS Number:  
Irritant
Zirconium(IV) oxide
Catalog # ROTH9303.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

About this item

 :
 :123.22 g/mol
Merck Index:14,10180
Boiling Pt:4300 °C (1013 hPa)
Melting Pt:2680 °C
Density:5.75 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-227-2
 :GHS07
CAS Number:1314-23-4
Formula:ZrO₂
MDL Number:MFCD00011310