
1,8-Naphthalenedicarboxylic anhydride 97%
:81-84-5
:
:C₁₂H₆O₃
:MFCD00006925
:425 °C (1013 hPa)
:>269 °C
:272 °C
:198.18 g/mol
:Ambient
:201-380-2
:GHS07
- Appearance:Beige to light brown Powder
- Infrared spectrum:Conforms
- Melting point:≥269 °C
- Loss on drying:≤0.5 %
- HPLC:≥96.0 %