Selenium trioxide, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: Selenium dioxide
- CAS Number:13768-86-0
- Formula:O₃Se
- Molecular Weight:126.96 g/mol
- Melting Pt:117.85…118.85 °C
- Density:3.6 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ200
- :Density: 3.6 g/cm³ (20 °C)
- EINECS:237-385-1
2 Options Available Below
Product Details & Documents
Synonyms: Selenium dioxide
- CAS Number:13768-86-0
- Formula:O₃Se
- Molecular Weight:126.96 g/mol
- Melting Pt:117.85…118.85 °C
- Density:3.6 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ200
- :Density: 3.6 g/cm³ (20 °C)
- EINECS:237-385-1



