Order Entry
Export
ContactUsLinkComponent
1,4-Bis(diphenylphosphino)butane ≥98%
Irritant
1,4-Bis(diphenylphosphino)butane ≥98%
Catalog # B21122.36
CAS Number:  
Irritant
1,4-Bis(diphenylphosphino)butane ≥98%
Catalog # B21122.36
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

About this item

 :
 :426.48 g/mol
Melting Pt:132…136 °C
Density:1.23 g/cm³ (20 °C)
EINECS:231-698-7
 :GHS07
Storage Temperature:Ambient
CAS Number:7688-25-7
Formula:C₂₈H₂₈P₂
MDL Number:MFCD00003051