
1,3-Bis(diphenylphosphino)propane 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6737-42-4
:
Formula:C₂₇H₂₆P₂
MDL Number:MFCD00003050
Melting Pt:57…63 °C
:412.45 g/mol
EINECS:229-791-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to yellow to light beige Powder or granules
- Infrared spectrum:Conforms
- Melting point:57 to 63 °C
- Elemental Analysis:≥96.0 %