Order Entry
Export
ContactUsLinkComponent
1,2-Bis(diphenylphosphino)ethane
Irritant
1,2-Bis(diphenylphosphino)ethane
Catalog # MOLEM18770139
Supplier:  Molekula
CAS Number:  
Irritant
1,2-Bis(diphenylphosphino)ethane
Catalog # MOLEM18770139
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :398.42 g/mol
Melting Pt:140…142 °C
EINECS:216-769-2
 :GHS07
Storage Temperature:Ambient
CAS Number:1663-45-2
Formula:C₂₆H₂₄P₂
MDL Number:MFCD00003047