Order Entry
Export
Orders LinkContactUsLinkComponent
 

Lead(II) oxide, Sigma-Aldrich®

Health
Irritant
Environment
Supplier:   Merck
Lead(II) oxide, Sigma-Aldrich®
Supplier:  Merck
Certificates
Lead(II) oxide, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
Synonyms:  Lead monoxide
  • CAS Number:1317-36-8
  • Formula:PbO
  • MDL Number:MFCD00011164
  • Molecular Weight:223.2 g/mol
  • Merck Index:13,05431
  • Boiling Pt:1470 °C (1013 hPa)
  • Melting Pt:888 °C
  • Density:9.5 g/cm³ (20 °C)
  • EINECS:215-267-0
  • ADR:6.1,III
  •  :M-Clarity™ quality level = MQ100
  • UN:2291
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 98,50 €
    98,50 €
    1 * 10 g
       98,50 €

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 396,00 €
    396,00 €
    1 * 50 g
       396,00 €