Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium hexametaphosphate
Sodium hexametaphosphate 65-70% (P₂O₅ basis), Sigma-Aldrich®
Sodium hexametaphosphate 65-70% (P₂O₅ basis), Sigma-Aldrich®
Sodium hexametaphosphate 65-70% (P₂O₅ basis), Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Sodium hexametaphosphate 65-70% (P₂O₅ basis), Sigma-Aldrich®
Supplier:
Merck
Certificates
About this item
CAS Number:
68915-31-1
:
10124-56-8
Formula:
(NaO₃P)n
MDL Number:
MFCD00134118
Melting Pt:
610 °C
Density:
1.25 g/cm³ (20 °C)
EINECS:
233-343-1
Related Information
Documentation
Certificates
Search by Lot Number
Product specification
SIAL71600-250G_TDS_EN
Documentation