

meta-Phosphoric acid 39 - 43% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:37267-86-0
:
Formula:(HPO₃)n
MDL Number:MFCD00011351
:79.98 g/mol
Merck Index:14,7345
Boiling Pt:260 °C (1013 hPa)
Melting Pt:21 °C
Density:2.2
UN:3260
ADR:8,III
:Stabilised with sodium metaphosphate
EINECS:253-433-4
:GHS05
:GHS07
Storage Temperature:Ambient