
4-Aminoantipyrine ≥98.0% analytical reagent
:83-07-8
:
:C₁₁H₁₃N₃O
:MFCD00003145
:13,00596
:107…109 °C
:203.24 g/mol
:201-452-3
:GHS07
:Ambient
- Assay:Min. 98.0 %
- Suitable as phenol reagent:Passes test
- Melting point:106 - 109 °C
- Loss on drying (90°C):Max. 0.5 %
- Ignition residue (SO4):Max. 0.1 %