Order Entry
Export
Orders LinkContactUsLinkComponent
PIPES, Sigma-Aldrich®

PIPES, Sigma-Aldrich®

Supplier:   Merck
PIPES, Sigma-Aldrich®
Supplier:  Merck
Certificates
PIPES, Sigma-Aldrich®
PIPES, Sigma-Aldrich®

4 Options Available Below

Product Details & Documents
Synonyms:  Piperazine-1,4-bis(2-ethanesulphonic acid), 2,2'-(Piperazine-1,4-diyl)diethanesulphonic acid, Piperazine-N,N'-bis-2-ethanesulphonic acid, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid
  • CAS Number:5625-37-6
  • Formula:C₈H₁₈N₂O₆S₂
  • MDL Number:MFCD00006159
  • Molecular Weight:302.37 g/mol
  • Merck Index:12,07633
  • Boiling Pt:499 °C (1013 hPa)
  • Melting Pt:300 °C
  •  :M-Clarity™ quality level = MQ400
  • Storage Temperature:Ambient
  • EINECS:227-057-6
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 99,20 €
    99,20 €
    1 * 25 g
       99,20 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 341,00 €
    341,00 €
    1 * 100 g
       341,00 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1 250,00 €
    1 250,00 €
    1 * 500 g
       1 250,00 €

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 1 990,00 €
    1 990,00 €
    1 * 1 kg
       1 990,00 €