PIPES 98.5+% for biochemistry
:5625-37-6
:
:C₈H₁₈N₂O₆S₂
:MFCD00006159
:302.37 g/mol
:12,07633
:499 °C (1013 hPa)
:>300 °C
:Ambient
:227-057-6
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 %
- UV:(0.1 M in 0.1 M NaOH) 1 cm cell
- Titration with NaOH:≥98.50 %
- Iron (Fe):≤15 ppm
- Lead (Pb):≤5 ppm