Order Entry
Export
Orders LinkContactUsLinkComponent
 

Diphenylcyclopropenone 98%

Diphenylcyclopropenone 98%
Certificates
Diphenylcyclopropenone 98%

3 Options Available Below

Product Details & Documents
Synonyms:  2,3-Diphenyl-2-cyclopropen-1-one, 2,3-Diphenylcycloprop-2-en-1-one, 2,3-Diphenyl-2-cyclopropen-1-on, 1,2-Diphenylcyclopropen-3-one
  • CAS Number:886-38-4
  • Formula:C₁₅H₁₀O
  • MDL Number:MFCD00001311
  • Molecular Weight:206.24 g/mol
  • Merck Index:12,03366
  • Boiling Pt:407 °C (1013 hPa)
  • Melting Pt:117…121 °C
  • Density:1,1 g/cm³ (20 °C)
  • Storage Temperature:Refrigerator
  • EINECS:212-948-4
Specifications for Product Family

Specification Test Results

  • Appearance:
    Beige to light orange-pink Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    117 to 121 °C
  • HPLC:
    ≥97.5 %
  • Microbial contamination:
    ≤5000 CFU/g (TAMC)
  • pH:
    6.5 to 7.5 (2 % in water)
  • Chlorides:
    ≤2 % (as NaCl)
  • Total nitrogen:
    9.3 to 10.7 %
  • Amino nitrogen:
    1.8 to 3.2 %
Product Family Options

Product Information