1,1-Diethoxycyclohexane, Sigma-Aldrich®
Catalog # SIAL539430-50ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 mL
- Seq:2370
Specifications
About this item
:
:172.27 g/mol
Boiling Pt:30…32 °C (0.4 mmHg)
Density:0.9309 g/cm³ (0 °C)
:M-Clarity™ quality level = MQ100
EINECS:216-798-0
CAS Number:1670-47-9
Formula:C₁₀H₂₀O₂
MDL Number:MFCD00037723