1,2-Epithiocyclohexane, Sigma-Aldrich®
Catalog # SIALC102601-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:114.21 g/mol
Boiling Pt:55 °C (1013 hPa)
Density:1.041 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:206-008-2
CAS Number:286-28-2
Formula:C₆H₁₀S
MDL Number:MFCD00005163