Tetrachloro-o-benzoquinone, Sigma-Aldrich®
SIALT6601-5G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
: 3,4,5,6-Tetrachloro-1,2-benzoquinone, Tetrachloro-1,2-benzoquinone, 2-Chloranil, Tetrachloro-o-benzoquinone, o-Chloranil, 3,4,5,6-Tetrachlorocyclohexa-3,5-diene-1,2-dione
:
:245.88 g/mol
:219-424-4
:2435-53-2
:C₆Cl₄O₂
:MFCD00001646
:312
:126…129 °C
:2.01 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100