Order Entry
Export
ContactUsLinkComponent
Tetrachloro-o-benzoquinone
Tetrachloro-o-benzoquinone
Catalog # MOLE73012947-25G
Supplier:  Molekula
CAS Number:  
Tetrachloro-o-benzoquinone
Catalog # MOLE73012947-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 3,4,5,6-Tetrachloro-1,2-benzoquinone, Tetrachloro-1,2-benzoquinone, 2-Chloranil, Tetrachloro-o-benzoquinone, o-Chloranil, 3,4,5,6-Tetrachlorocyclohexa-3,5-diene-1,2-dione
 :
 :245.88 g/mol
EINECS:219-424-4
CAS Number:2435-53-2
Formula:C₆Cl₄O₂
MDL Number:MFCD00001646
Boiling Pt:312
Melting Pt:126…129 °C
Density:2.01 g/cm³ (20 °C)