2,3-Dichloro-5,6-dicyano-p-benzoquinone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:84-58-2
:
Formula:C₈Cl₂N₂O₂
MDL Number:MFCD00001593
:227.01 g/mol
Merck Index:13,03088
Boiling Pt:396 °C (1013 hPa)
Melting Pt:209…214 °C
Density:1.713 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:M-Clarity™ quality level = MQ200
EINECS:201-542-2