Order Entry
Export
ContactUsLinkComponent
 
 
1-Phenyl-1,2-propanedione 98%
 
undefined
 
1-Phenyl-1,2-propanedione 98%

 

  • Appearance:
    Clear yellow to orange Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5300 to 1.5340 (20°C, 589 nm)
  • GC:
    ≥97.5 %