Order Entry
Export
Orders LinkContactUsLinkComponent
2'-Hydroxy-4'-methoxyacetophenone, Sigma-Aldrich®

2'-Hydroxy-4'-methoxyacetophenone, Sigma-Aldrich®

Supplier:   Merck
2'-Hydroxy-4'-methoxyacetophenone, Sigma-Aldrich®
Supplier:  Merck
Certificates
2'-Hydroxy-4'-methoxyacetophenone, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
Synonyms:  1-(2-Hydroxy-4-methoxyphenyl)ethanone, Paeonol
  • CAS Number:552-41-0
  • Formula:C₉H₁₀O₃
  • MDL Number:MFCD00008730
  • Molecular Weight:166.18 g/mol
  • Boiling Pt:154 °C (20 mmHg)
  • Melting Pt:51 °C
  • Density:1.1604 g/cm³ (40 °C)
  •  :M-Clarity™ quality level = MQ100
  • EINECS:209-012-2
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 28,00 €
    28,00 €
    1 * 1 g
       28,00 €

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 96,10 €
    96,10 €
    1 * 10 g
       96,10 €