

2,6-Dibromo-p-toluidine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6968-24-7
:
Formula:C₇H₇Br₂N
MDL Number:MFCD00007641
Boiling Pt:296 °C (1013 hPa)
Melting Pt:72…76 °C
:264.95 g/mol
EINECS:230-182-9
:GHS06
:GHS07
Storage Temperature:Ambient