Order Entry
Export
ContactUsLinkComponent
2,4'-DDE
Flammable
Toxic
2,4'-DDE
Catalog # EHERC12040000
Supplier:  EHRENSTORFER
CAS Number:  
Flammable
Toxic
2,4'-DDE
Catalog # EHERC12040000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :318.03 g/mol
Boiling Pt:64…65 °C (1013 hPa)
Melting Pt:–98 °C
Flash Pt:11 °C
Storage Temperature:Refrigerator
EINECS:222-318-0
UN:2930
ADR:6.1,II
 :GHS02
 :GHS06
CAS Number:3424-82-6
Formula:C₁₄H₈Cl₄