1,3-Dinitro-4-(trifluoromethoxy)benzene
Catalog # MOLE37605955-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:252.11 g/mol
EINECS:470-480-8
UN:2927
ADR:6.1,II
CAS Number:655-07-2
Formula:C₇H₃F₃N₂O₅
MDL Number:MFCD00042268