Order Entry
Export
ContactUsLinkComponent
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB), Molecular Biology Grade
Irritant
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB), Molecular Biology Grade
Catalog # BC87
Supplier:  G-Biosciences
CAS Number:  
Irritant
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB), Molecular Biology Grade
Catalog # BC87
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :396.36 g/mol
Melting Pt:240…244 °C
EINECS:200-714-4
 :GHS07
Storage Temperature:Refrigerator
CAS Number:69-78-3
Formula:C₁₄H₈N₂O₈S₂
MDL Number:MFCD00007140