Order Entry
Export
ContactUsLinkComponent
 
Propyl-3,4,5-trihydroxybenzoate for synthesis, Sigma-Aldrich®
 :  Merck
 
Propyl-3,4,5-trihydroxybenzoate for synthesis, Sigma-Aldrich®
 :  Merck
 

 

  • Environmentally Preferable:

 

 

  • Solubility:
    3.5 g/l (25 °C)
  • Melting Point:
    145 - 148 °C
  • Molar Mass:
    212.2 g/mol
  • Flash Point:
    187 °C
  • Synonyms:
    Propyl gallate, Gallic acid propyl ester, 3,4,5-Trihydroxybenzoic acid propyl ester
  • Hill Formula:
    C10H12O5
  • Chemical Formula:
    3,4,5-(OH)3C6H2COOCH2CH2CH3