Order Entry
Export
Orders LinkContactUsLinkComponent
 

Ethyl-3,4,5-trihydroxybenzoate

Glass bottle for solids, 25 g
Catalog # ROTH3629.1
Supplier:   Roth Carl
Ethyl-3,4,5-trihydroxybenzoate
Catalog # ROTH3629.1
Supplier:  Roth Carl
CAS Number:  831-61-8
Ethyl-3,4,5-trihydroxybenzoate

Glass bottle for solids, 25 g

CAS Number:  831-61-8
Price
45,60 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Gallic acid ethyl ester, Ethyl 3,4,5-trihydroxybenzoate, Ethyl gallate
  • CAS Number:831-61-8
  • Formula:C₉H₁₀O₅
  • MDL Number:MFCD00016430
  • Molecular Weight:198.18 g/mol
  • EINECS:212-608-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications