MOPS-Na, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:71119-22-7
:
Formula:C₇H₁₄NNaO₄S
MDL Number:MFCD00064350
:231.25 g/mol
Storage Temperature:Ambient
EINECS:428-420-3
Specification Test Results
- pH:9,0 - 10,5 (1% aq solution)
- pka:pKa −7,2 at 25 °C
- Water Content:≤0,5%