Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
γ-(N-Morpholino)propanesulphonic acid (MOPS)
γ-(N-Morpholino)propanesulphonic acid (MOPS)
γ-(N-Morpholino)propanesulphonic acid (MOPS)
:
PanReac AppliChem
γ-(N-Morpholino)propanesulphonic acid (MOPS)
:
PanReac AppliChem
:
1132-61-2
:
:
C₇H₁₅NO₄S
:
MFCD00006183
:
209.27 g/mol
:
13,06288
:
> 250 °C
:
110 °C (closed cup)
:
Ambient
:
214-478-5