Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Toluene-α-sulphonyl fluoride
Toluene-α-sulphonyl fluoride, Sigma-Aldrich®
Toluene-α-sulphonyl fluoride, Sigma-Aldrich®
:
Merck
Toluene-α-sulphonyl fluoride, Sigma-Aldrich®
:
Merck
:
329-98-6
:
:
C₇H₇FO₂S
:
MFCD00007424
:
174.2 g/mol
:
80…85 °C (0.3 mmHg)
:
91…92 °C
:
Ambient
:
206-350-2
:
2923
:
8,III
:
GHS06
:
GHS05