Order Entry
Export
ContactUsLinkComponent
Toluene-α-sulphonyl fluoride
Toxic
Corrosive
Toluene-α-sulphonyl fluoride
Catalog # ROCH10837091001
Supplier:  ROCHE
CAS Number:  
Toxic
Corrosive
Toluene-α-sulphonyl fluoride
Catalog # ROCH10837091001
Supplier:  ROCHE
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :174.2 g/mol
Boiling Pt:80…85 °C (0.3 mmHg)
Melting Pt:91…92 °C
ADR:8,III
 :M-Clarity™ quality level = MQ100
EINECS:206-350-2
UN:2923
 :GHS06
 :GHS05
Storage Temperature:Ambient
CAS Number:329-98-6
Formula:C₇H₇FO₂S
MDL Number:MFCD00007424