
Heptadecanedioic acid ≥95.0% (by titrimetric analysis)
Catalog # TCIAH0972-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 90.0 %
- Purity (Neutralization titration):min. 95.0 %
- Melting point:117.0 to 121.0 deg-C
Specification Test Results
About this item
:
:300.44 g/mol
:GHS07
CAS Number:2424-90-0
Formula:C₁₇H₃₂O₄
MDL Number:MFCD00673439